missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Bcl XL Protein

A cDNA sequence encoding the Bcl XL was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP11140-1mg

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15916278

  • 2610.00€ / 1mg
Expédition estimée: 24-05-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Spécification

Spécification

Bcl XL Protein
Research Use Only
Recombinant Protein
E. coli
SQSNRELVVD FLSYKLSQKG YSWSQFSDVE ENRTEAPEET EAERETPSAI NGNPSWHLAD SPAVNGATGH SSSLDAREVI PMAAVKQALR EAGDEFELRY RRAFSDLTSQ LHITPGTAYQ SFEQVVNELF RDGVNWGRIV AFFSFGGALC VESVDKEMQV LVSRIASWMA TYLNDHLEPW IQENGGWDTF VDLYGNNAAA ESRKGQERFN R
Greater than 97.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Mouse
Untagged
Bcl-XL Mouse was lyophilized from a 0.2microm filtered concentrated solution in 1M PBS, pH 7.4 and 5% Trehalose.
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions