missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Cd163 Protein
A DNA sequence encoding the Mus musculus (Mouse) Cd163, was expressed in the hosts and tags indicated.
Marque: enQuireBio™ QP7641-ye-500ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
NP_444324.2 | |
Cd163 Protein | |
Research Use Only | |
Cd163 | |
Recombinant Protein | |
Yeast | |
VVCQQLGCPT SIKALGWANS SAGSGYIWMD KVSCTGNESA LWDCKHDGWG KHNCTHEKDA GVTCSDGSNL EMRLVNSAGH RCLGRVEIKF QGKWGTVCDD NFSKDHASVI CKQLGCGSAI SFSGSAKLGA GSGPIWLDDL ACNGNESALW DCKHRGWGKH NCDHAEDVGV ICLEGADLSL RLVDGVSRCS GRLEVRFQGE WGTVCDDNWD LRDASVVCKQ LGCPTAISAI GRVNASEGSG QIWLDNISCE GHEATLWECK HQEWGKHYCH HREDAGVTCS | |
Greater than 90% as determined by SDS-PAGE. |
93671 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |