missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse CD55 Protein

A cDNA sequence encoding the CD55 was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP11339-10ug

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15996918

  • 203.00€ / 10µg
Expédition estimée: 07-06-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Spécification

Spécification

13136
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
Baculovirus
DCGPPPDIPN ARPILGRHSK FAEQSKVAYS CNNGFKQVPD KSNIVVCLEN GQWSSHETFC EKSCVAPERL SFASLKKEYL NMNFFPVGTI VEYECRPGFR KQPPLPGKAT CLEDLVWSPV AQFCKKKSCP NPKDLDNGHI NIPTGILFGS EINFSCNPGY RLVGVSSTFC SVTGNTVDWD DEFPVCTEIH CPEPPKINNG IMRGESDSYT YSQVVTYSCD KGFILVGNAS IYCTVSKSDV GQWSSPPPRC IEKSKVPTKK PTINVPSTGT PSTPQKPTTE SVPNPGDQPT PQKPSTVKVS ATQHVPVTKT TVRHPIRTST DKGEPNTGLE HHHHHH
Greater than 95.0% as determined by SDS-PAGE.
CD55 Protein
Research Use Only
Cd55
Mouse
His
CD55 protein solution (0.5 mg/ml) contains Phosphate Buffered Saline (pH 7.4) and 10% glycerol.
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions