missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Clathrin light chain A Protein
A DNA sequence encoding the Mus musculus (Mouse) Clathrin light chain A, was expressed in the hosts and tags indicated.
Marque: enQuireBio™ QP5841-ec-500ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
Clathrin light chain A Protein | |
Research Use Only | |
Clta | |
Recombinant Protein | |
E. coli | |
MAELDPFGAP AGAPGGPALG NGVAGAGEED PAAAFLAQQE SEIAGIENDE AFAILDGGAP GRATRRAAGG PDAVDGVMNG EYYQESNGPT DSYAAISEVD RLQSEPESIR KWREEQTERL EALDANSRKQ EAEWKEKAIK ELEEWYARQD EQLQKTKANN RVADEAFYKQ PFADLIGYVA AEEAFVNDID ESSPGTEWER VARLCDFNPK SSKQAKDVSR MRSVLISLKQ APLVH | |
Greater than 90% as determined by SDS-PAGE. |
500 μg | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |