missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse complement factor B Protein
A DNA sequence encoding the Mus musculus (Mouse) complement factor B, was expressed in the hosts and tags indicated.
Marque: enQuireBio™ QP5822-ec-100ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
NP_001701.2 | |
complement factor B Protein | |
Research Use Only | |
CFB | |
Recombinant Protein | |
E. coli | |
VTTTPWSLAR PQGSCSLEGV EIKGGSFRLL QEGQALEYVC PSGFYPYPVQ TRTCRSTGSW STLKTQDQKT VRKAECRAIH CPRPHDFENG EYWPRSPYYN VSDEISFHCY DGYTLRGSAN RTCQVNGRWS GQTAICDNGA GYCSNPGIPI GTRKVGSQYR LEDSVTYHCS RGLTLRGSQR RTCQEGGSWS GTEPSCQDSF MYDTPQEVAE AFLSSLTETI EGVDAEDGHG PGEQQKRKIV LDPSGSMNIY LVLDGSDSIG ASNFTGAKKC LVNLIEKVAS YGVKPRYGLV TYATYPKIWV KVSEADSSNA DWVTKQLNEI NYEDHKLKSG TNTKKALQAV YSMMSWPDDV PPEGWNRTRH VIILMTDGLH NMGGDPITVI DEIRDLLYIG KDRKNPREDY LDVYVFGVGP LVNQVNINAL ASKKDNEQHV FKVKDMENLE DVFYQMIDES QSLSLCGMVW EHRKGTDYHK QPWQAKISVI RPSKGHESCM GAVVSEYFVL TAAHCFTVDD KEHSIKVSVG GEKRDLEIEV VLFHPNYNIN GKKEAGIPEF YDYDVALIKL KNKLKYGQTI RPICLPCTEG TTRALRLPPT TTCQQQKEEL LPAQDIKALF VSEEEKKLTR KEVYIKNGDK KGSCERDAQY APGYDKVKDI SEVVTPRFLC TGGVSPYADP NTCRGDSGGP LIVHKRSRFI QVGVISWGVV DVCKNQKRQK QVPAHARDFH INLFQVLPWL KEKLQDEDL | |
Greater than 90% as determined by SDS-PAGE. |
629 | |
100 μg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |