Learn More
enQuireBio™ Recombinant Mouse ENO2 Protein
A cDNA sequence encoding the ENO2 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP10589-100ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
13807 | |
Greater than 95% as determined by SDS-PAGE. | |
Research Use Only | |
Eno2 | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMSIEKIW AREILDSRGN PTVEVDLYTA KGLFRAAVPS GASTGIYEAL ELRDGDKQRY LGKGVLKAVD HINSRIAPAL ISSGISVVEQ EKLDNLMLEL DGTENKSKFG ANAILGVSLA VCKAGAAERD LPLYRHIAQL AGNSDLILPV PAFNVINGGS HAGNKLAMQE FMILPVGAES FRDAMRLGAE VYHTLKGVIK DKYGKDATNV GDEGGFAPNI LENSEALELV KEAIDKAGYT EKMVIGMDVA ASEFYRDGKY DLDFKSPADP SRYITGDQLG ALYQDFVRNY PVVSIEDPFD QDDWAAWSKF TANVGIQIVG DDLTVTNPKR IERAVEEKAC NCLLLKVNQI GSVTEAIQAC KLAQENGWGV MVSHRSGETE DTFIADLVVG LCTGQIKTGA PCRSERLAKY NQLMRIEEEL GDEARFAGHN FRNPSVL |
ENO2 Protein | |
100 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Specific activity is > 10,000 pmol/min/microg, and was obtained by measuring the decrease of NAD in absorbance at 340nm resulting from NADH at pH 6.5 at 37C. | |
Mouse | |
His | |
The ENO2 solution (1 mg/ml) contains Phosphate Buffered Saline (pH 7.4) and 10% glycerol. |