missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse FGF9 Protein
Recombinant Protein
Marque: enQuireBio™ QP5414-1mg
Informations supplémentaires : Poids : 0.01000kg
Spécification
NP_038546.2 | |
FGF9 Protein | |
1 mg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Mouse FGF9 induces 3T3 cell proliferation (typical ED50 is ? 2 ng/ml). | |
Mouse | |
Untagged | |
10 mM sodium phosphate, pH 7.5 |
14180 | |
Greater than 95% as determined by SDS-PAGE. | |
Research Use Only | |
FGF9 | |
Recombinant Protein | |
E. coli | |
MPLGEVGSYF GVQDAVPFGN VPVLPVDSPV LLNDHLGQSE AGGLPRGPAV TDLDHLKGIL RRRQLYCRTG FHLEIFPNGT IQGTRKDHSR FGILEFISIA VGLVSIRGVD SGLYLGMNEK GELYGSEKLT QECVFREQFE ENWYNTYSSN LYKHVDTGRR YYVALNKDGT PREGTRTKRH QKFTHFLPRP VDPDKVPELY KDILSQS |