Learn More
enQuireBio™ Recombinant Mouse IL 28A Protein
A cDNA sequence encoding the IL 28A was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP10708-5ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
330496 | |
Greater than 95% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. | |
Research Use Only | |
Ifnl2 | |
Recombinant Protein | |
E. coli | |
DPVPRATRLP VEAKDCHIAQ FKSLSPKELQ AFKKAKDAIE KRLLEKDMRC SSHLISRAWD LKQLQVQERP KALQAEVALT LKVWENMTDS ALATILGQPL HTLSHIHSQL QTCTQLQATA EPKPPSRRLS RWLHRLQEAQ SKETPGCLED SVTSNLFRLL TRDLKCVASG DQCV |
IL 28A Protein | |
5 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
The ED50 as determined by an anti-viral assay using human HepG2 cells infected with encephalomyocarditis(EMC) virus is less than 3 ng/ml, corresponding to a specific activity of > 3.3 M 105 IU/mg. | |
Mouse | |
Untagged | |
The Mouse IL-28A is lyophilized from a 0.2microm filtered concentrated solution in PBS, pH 7.4 and 5% trehalose. |