Learn More
enQuireBio™ Recombinant Mouse IL16 / Interleukin-16 Protein
Recombinant Protein
Marque:  enQuireBio™ QP5456-500ug
Informations supplémentaires : Poids : 0.01000kg
Code produit. 15926169
  • 4305.00€ / 500µg
Expédition estimée: 30-06-2022
Spécification
NP_034681.2
IL16 / Interleukin-16 Protein
500 μg
Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Mouse IL16 / Interleukin-16 induces primary human T cell chemotaxis (significanty different from control at Mouse IL16 / Interleukin-16 concentrations greater than 5 ng/ml).
Mouse
Untagged
10 mM sodium phosphate, pH 7.5
16170
Greater than 95% as determined by SDS-PAGE.
Research Use Only
IL16
Recombinant Protein
E. coli
MHDLNSSTDS AASASAASDI SVESKEATVC TVTLEKTSAG LGFSLEGGKG SLHGDKPLTI NRIFKGDRTG EMVQPGDEIL QLAGTAVQGL TRFEAWNVIK ALPDGPVTIV IRRTSLQCKQ TTASADS