missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse IL25 Protein
Recombinant Protein
Marque: enQuireBio™ QP5467-500ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
IL25 Protein | |
500 μg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Mouse IL25 bioactivity was confirmed in a PBMC IL-8 production assay (see figure). | |
Mouse | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |
Greater than 90 as determined by SDS-PAGE. | |
Research Use Only | |
IL25 | |
Recombinant Protein | |
E. coli | |
MVSLRIQEGC SHLPSCCPSK EQEPPEEWLK WSSASVSPPE PLSHTHHAES CRASKDGPLN SRAISPWSYE LDRDLNRVPQ DLYHARCLCP HCVSLQTGSH MDPLGNSVPL YHNQTVFYRR PCHGEEGTHR RYCLERRLYR VSLACVCVRP RVMA |