missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Noggin Protein

A cDNA sequence encoding the Noggin was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP10795-20ug

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15905198

  • 215.00€ / 20µg
Expédition estimée: 10-05-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Spécification

Spécification

Noggin Protein
Research Use Only
The ED50 as determined by inhibiting BMP-4-induced alkaline phosphatase production of murine ATDC5 cells is less than 2ng/ml, corresponding to a specific activity of > 5.0 M 105 IU/mg in the presence of 5ng/ml BMP-4.
Mouse
Untagged
Lyophilized from a 0.2?m filtered solution in 30% acetonitrile, 0.1% TFA.
20 μg
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
E. coli
MQHYLHIRPAPSDNLPLVDLIEHPDPIFDPKEKDLNETLLRSLLGGHYDPGFMATSPPEDRPGGGGGPAGGAEDLAELDQLLRQRPSGAMPSEIKGLEFSEGLAQGKKQRLSKKLRRKLQMWLWSQTFCPVLYAWNDLGSRFWPRYVKVGSCFSKRSCSVPEGMVCKPSKSVHLTVLRWRCQRRGQRCGWIPIQYPIISECKCSC
Greater than 95.0% as determined by SDS-PAGE.
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions