missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse OSM Protein

A cDNA sequence encoding the OSM was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP10801-2ug

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15995208

  • 137.00€ / 2µg
Expédition estimée: 04-08-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Spécification

Spécification

OSM Protein
2 μg
< 1.0 EU per ug protein as determined by the LAL method.
Mouse
Untagged
OSM protein was lyophilized from a 0.2microm filtered concentrated solution in 1xPBS, pH 7.4.
Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
NRGCSNSSSQ LLSQLQNQAN LTGNTESLLE PYIRLQNLNT PDLRAACTQH SVAFPSEDTL RQLSKPHFLS TVYTTLDRVL YQLDALRQKF LKTPAFPKLD SARHNILGIR NNVFCMARLL NHSLEIPEPT QTDSGASRST TTPDVFNTKI GSCGFLWGYH RFMGSVGRVF REWDDGSTRS R
Vidéos
FDS
Documents
Certificats
Promotions

Promotions