Learn More
enQuireBio™ Recombinant Mouse S100A1 Protein
A cDNA sequence encoding the S100A1 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP13386-5ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
S100A1 Protein | |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Mouse | |
His | |
S100A1 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 30% glycerol and 0.1M NaCl. |
Greater than 90.0% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSELESAME TLINVFHAHS GKEGDKYKLS KKELKDLLQT ELSGFLDVQK DADAVDKVMK ELDENGDGEV DFKEYVVLVA ALTVACNNFF WETS |