missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse SAMHD1 Protein

A DNA sequence encoding the Mus musculus (Mouse) SAMHD1, was expressed in the hosts and tags indicated.

Marque:  enQuireBio™ QP6650-ec-200ug

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 16034820

Cet article est actuellement indisponible ou discontinu. Veuillez consulter la page produit pour de possible alternative dans l’onglet


Description

Description

Spécification

Spécification

NP_001132992.1
SAMHD1 Protein
Research Use Only
SAMHD1
Recombinant Protein
E. coli
DIMITDAFLK ADPYVEITGT AGKKFRISTA IDDMEAFTKL TDNIFLEVLH STDPQLSEAQ SILRNIECRN LYKYLGETQP KREKIRKEEY ERLPQEVAKA KPEKAPDVEL KAEDFIVDVI NVDYGMEDKN PIDRVHFYCK SNSKQAVRIN KEQVSQLLPE KFAEQLIRVY CKKKDGKSLD AAGKHFVQWC ALRDFTKPQD GDIIAPLITP LKWNNKTSSC LQEVSKVKTC LK
Greater than 90% as determined by SDS-PAGE.
56045
200 μg
Not determined.
Partial (see sequence information for more details).
Mouse
His
Tris-based buffer, 50% glycerol
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions