Learn More
enQuireBio™ Recombinant Mouse UCHL1 Protein
A cDNA sequence encoding the UCHL1 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP10904-10ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
UCHL1 Protein | |
10 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMQLKPME INPEMLNKVL AKLGVAGQWR FADVLGLEEE TLGSVPSPAC ALLLLFPLTA QHENFRKKQI EELKGQEVSP KVYFMKQTIG NSCGTIGLIH AVANNQDKLE FEDGSVLKQF LSETEKLSPE DRAKCFEKNE AIQAAHDSVA QEGQCRVDDK VNFHFILFNN VDGHLYELDG RMPFPVNHGA SSEDSLLQDA AKVCREFTER EQGEVRFSAV ALCKAA |
Greater than 90.0% as determined by SDS-PAGE. | |
Research Use Only | |
Specific activity is > 70 pmol/min/ug, and is defined as the amount of enzyme that hydrolysis 1.0 pmole of ubiquitin-AMC per minute at pH 7.5, at 37C. | |
Mouse | |
His | |
UCHL1 protein solution (1 mg/ml) containing Phosphate buffered saline (pH 7.4), 10% glycerol and 1mM DTT. |