Learn More
enQuireBio™ Recombinant other AsnRS Protein
A cDNA sequence encoding the AsnRS was constructed and used to recombinantly synthesize the protein.
Marque:  enQuireBio™ QP11088-50ug
Informations supplémentaires : Poids : 0.01000kg
Code produit. 15916108
  • 203.00€ / 50µg
Expédition estimée: 10-05-2024
Spécification
AsnRS Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MTVYICPETG DDGNDGSELK PLRTLYQAMI ITKSSKGDFL IRTKKDGKQV WEAASKTALK KSWKRYEQEM LKNEKVAAKM LEKDATEVGV KAALEEAKKV QIELDTSLSY ITGVKIRDLV KHRNERVCIK GWIHRMRRQG KSLMFFILRD GTGFLQVLLM DKLCQTYDAL TVNTECTVEI YGAIKEVPEG KEAPNGHELI ADFWKIIGNA PSGGIDNVLN EEASVDKMLD NRHLVIRGEN AAALLRLRAA ATRAMREHFY NAGYVEVAPP TLVQTQVEGG STLFNLDYFG EQSFLTQSSQ LYLETCIPTL GDVFLHCSVL QGGKISHSST LAEYAHVEAE CPFITLDDLM EKIEELVCDT VDRLLADEEA KKLLEHINPK FQPPERPFLR MEYKDAIKWL QEHNVENEFG NTFTYGEDIA EAAERFMTDT INKPILLNRF PSEIKAFYMQ RDAKDNTLTE SVDLLMPGVG EIVGGSMRIW KFDELSKAFK NVEIDPKPYY WYLDQRLYGT CPHGGYGLGL ERFICWLTNT NHIRDVCLYP RFVGRCVP
Greater than 95.0% as determined by SDS-PAGE.
50 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Other
His
AsnRS 0.2microm filtered solution containing 20mM HEPES, pH 7.4, 100mM NaCl, 5mM MgCl2, 5mM b-Mercaptoethanol and 10 % Glycerol.