missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other Filamin Protein

A cDNA sequence encoding the Filamin was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP11883-10ug

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15998678

  • 137.00€ / 10µg
Expédition estimée: 09-08-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Spécification

Spécification

Filamin Protein
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Other
Untagged
Greater than 90.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
Native Protein
The protein was lyophilized from a 1 mg/ml solution containing 20mM Tris / acetate buffer pH 7.6, 0.1mM EDTA, 2mM DTT, 9M urea and 20mM NaCl.
Vidéos
FDS
Documents
Certificats
Promotions

Promotions