Learn More
enQuireBio™ Recombinant other LLO PEST free Protein
A cDNA sequence encoding the LLO PEST free was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP10774-50ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
LLO PEST free Protein | |
50 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
7x104 HU/mg. 2mM DTT could be use to reactivate the toxin. | |
Other | |
Untagged | |
The protein contains 50mM NaH2PO4, 1mM EDTA, 2.7mM KCl, 1mM DTT, 5% glycerol and 0.5M NaCl. |
Greater than 90.0% as determined by SDS-PAGE. | |
Research Use Only | |
hlyA | |
Recombinant Protein | |
E. coli | |
MKKIMLVFIT LILISLPIAQ QTEAKDASAF HKEDLISSMA PPTSPPASPK TPIEKKHADE IDKYIQGLDY NKNNVLVYHG DAVTNVPPRK GYKDGNEYIV VEKKKKSINQ NNADIQVVNA ISSLTYPGAL VKANSELVEN QPDVLPVKRD SLTLSIDLPG MTNQDNKIVV KNATKSNVNN AVNTLVERWN EKYAQAYPNV SAKIDYDDEM AYSESQLIAK FGTAFKAVNN SLNVNFGAIS EGKMQEEVIS FKQIYYNVNV NEPTRPSRFF GKAVTKEQLQ ALGVNAENPP AYISSVAYGR QVYLKLSTNS HSTKVKAAFD AAVSGKSVSG DVELTNIIKN SSFKAVIYGG SAKDEVQIID GNLGDLRDIL KKGATFNRET PGVPIAYTTN FLKDNELAVI KNNSEYIETT SKAYTDGKIN IDHSGGYVAQ FNISWDEINY DPEGNEIVQH KNWSENNKSK LAHFTSSIYL PGNARNINVY AKECTGLAWE WWRTVIDDRN LPLVKNRNIS IWGTTLYPKY SNSVDNPIE |