Learn More
enQuireBio™ Recombinant other UNG Protein
A cDNA sequence encoding the UNG was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP13896-2,000U
Informations supplémentaires : Poids : 0.01000kg
Spécification
UNG Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
200mM Tris-HCl (pH 8.0 at 25C), 10mM DTT and 10mM EDTA. |
2000 IU | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
Untagged |