missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other Y. Enterocolitica (O:9) YopD Protein

A cDNA sequence encoding the Y. Enterocolitica (O:9) YopD was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP13976-50ug

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15905299

  • 870.00€ / 50µg
Expédition estimée: 10-05-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Spécification

Spécification

Y. Enterocolitica (O:9) YopD Protein
Research Use Only
Recombinant Protein
E. coli
Y. Enterocolitica (O:9) YopD is supplied in 20mM HEPES buffer pH 7.6, 250mM NaCl and 20% glycerol.
50 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Other
His
Greater than 80.0% as determined by SDS-PAGE.
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions