Learn More
enQuireBio™ Recombinant P. aeruginosa Beta-lactamase OXA-10 Protein
A DNA sequence encoding the Pseudomonas aeruginosa Beta-lactamase OXA-10, was expressed in the hosts and tags indicated.
Marque: enQuireBio™ QP7059-1mg
Informations supplémentaires : Poids : 0.01000kg
Spécification
WP_000846390.1 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
bla | |
Recombinant Protein | |
E. coli | |
SITENTSWNK EFSAEAVNGV FVLCKSSSKS CATNDLARAS KEYLPASTFK IPNAIIGLET GVIKNEHQVF KWDGKPRAMK QWERDLTLRG AIQVSAVPVF QQIAREVGEV RMQKYLKKFS YGNQNISGGI DKFWLEGQLR ISAVNQVEFL ESLYLNKLSA SKENQLIVKE ALVTEAAPEY LVHSKTGFSG VGTESNPGVA WWVGWVEKET EVYFFAFNMD IDNESKLPLR KSIPTKIMES EGIIGG |
P. aeruginosa Beta-lactamase OXA-10 Protein | |
1 mg | |
Not determined. | |
Full Length | |
P. aeruginosa | |
His-SUMO | |
Tris-based buffer, 50% glycerol |