Learn More
enQuireBio™ Recombinant Rabbit IGF1 Protein
A cDNA sequence encoding the IGF1 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP10679-1mg
Informations supplémentaires : Poids : 0.01000kg
Spécification
IGF1 Protein | |
1 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MTAGPETLCG AELVDALQFV CGDRGFYFNK PTGYGSSSRR APQTGIVDEC CFRSCDLRRL EMYCAPLKP |
Greater than 98.0% as determined by:(a) Gel-filtration chromatography under non denaturing conditions. (b) Analysis by SDS-PAGE. | |
Research Use Only | |
IGF-I is biologically active when compared to human IGF-1. The ED50, calculated by the dose -dependent proliferation of human MCF/7 cells is 5 to 25ng/ml in the cell culture mixture dependent on culture conditions. Its activity consists of 30-40 % compared to human IGF-1. | |
Rabbit | |
Untagged | |
The protein was lyophilized from a concentrated (0.5 mg/ml) solution with 0.02% NaHCO3. |