Learn More
enQuireBio™ Recombinant Rat IL-17AF Protein
Recombinant Protein
Marque:  enQuireBio™ QP5206-1mg
Informations supplémentaires : Poids : 0.01000kg
Code produit. 15995669
  • 7360.00€ / 1mg
Expédition estimée: 04-08-2022
Spécification
IL-17AF Protein
1 mg
Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Rat IL-17AF induces IL-6 production in 3T3 cells as detected via ELISA (each lot is tested and typical ED50 is less than 20 ng/ml).
Rat
Untagged
Sterile filtered (0.22 um) solution containing 0.1% TFA
Greater than 95% as determined by SDS-PAGE.
Research Use Only
IL17
Recombinant Protein
E. coli
IL-17A:MAV LIPQSSVCPN AEANNFLQNV KVNLKVLNSL SSKASSRRPS DYLNRSTSPW TLSRNEDPDR YPSVIWEAQC RHQRCVNAEG KLDHHMNSVL IQQEILVLKR EPEKCPFTFR VEKMLVGVGC TCVSSIVRHA S IL-17F:MA RRNPKVGLSA LQKAGNCPPL EDNSVRVDIR IFNQNQGISV PRDFQNRSSS PWDYNITRDP DRFPSEIAEA QCRHSGCINA QGQEDGSMNS VPIQQEILVL RREPQGCSNS FRLEKMLIKV GCTCVTPIVH HAA