Learn More
enQuireBio™ Recombinant Rat ITGB1 / Integrin beta-1 / CD29 Protein
A DNA sequence encoding the Rattus norvegicus (Rat) ITGB1 / Integrin beta-1 / CD29, was expressed in the hosts and tags indicated.
Marque: enQuireBio™ QP6242-ec-1mg
Informations supplémentaires : Poids : 0.01000kg
Spécification
NP_058718.2 | |
ITGB1 / Integrin beta-1 / CD29 Protein | |
1 mg | |
Not determined. | |
Extracellular Domain | |
Rat | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
24511 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
Itgb1 | |
Recombinant Protein | |
E. coli | |
QTDKNRCLKA NAKSCGECIQ AGPNCGWCTN TTFLQEGMPT SARCDDLEAL KKKGCHPSDI ENPRGSQTIK KNKNVTNRSK GMAEKLRPED ITQIQPQQLL LKLRSGEPQK FTLKFKRAED YPIDLYYLMD LSYSMKDDLE NVKSLGTDLM NEMRRITSDF RIGFGSFVEK TVMPYISTTP AKLRNPCTSE QNCTSPFSYK NVLSLTDRGE FFNELVGQQR ISGNLDSPEG GFDAIMQVAV CGSLIGWRNV TRLLVFSTDA GFHFAGDGKL GGIVLPNDGQ CHLENNVYTM SHYYDYPSIA HLVQKLSENN IQTIFAVTEE FQPVYKELKN LIPKSAVGTL SGNSSNVIQL IIDAYNSLSS EVILENSKLP DGVTINYKSY CKNGVNGTGE NGRKCSNISI GDEVQFEISI TANKCPNKES ENQLKLNPLG FTEEVEVVLQ FICKCNCQSH GIPASPKCHE GNGTFECGAC RCNEGRVGRH CECSTDEVNS EDMDAYCRKE NSSEICSNNG ECVCGQCVCR KRENTNEIYS GKFCECDNFN CDRSNGLICG GNGVCRCRVC ECYPNYTGSA CDCSLDTVPC VATNGQICNG RGICECGACK CTDPKFQGPT CETCQTCLGV CAEHKECVQC RAFNKGEKKD TCAQECSHFN LTKVESREKL PQPVQVDPVT HCKEKDIDDC WFYFTYSVNS KGEAHVHVVE TPDCPTGPD |