missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat OSM Protein
A cDNA sequence encoding the OSM was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP10802-10ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
OSM Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
KRGCSSSSPK LLSQLKSQAN ITGNTASLLE PYILHQNLNT LTLRAACTEH PVAFPSEDML RQLSKPDFLS TVHATLGRVW HQLGAFRQQF PKIQDFPELE RARQNIQGIR NNVYCMARLL HPPLEIPEPT QADSGTSRPT TTAPGIFQIK IDSCRFLWGY HRFMGSVGRV FEEWGDGSRR SR RHSPLWAW LKGDHRIRPS RSSQSAMLRS LVPR | |
Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
10 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Rat | |
Untagged | |
OSM protein was lyophilized from a 0.2microm filtered concentrated solution in 1xPBS, pH 7.4. |