missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rhesus Macaque Flt3-Ligand Protein

A cDNA sequence encoding the Flt3-Ligand was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP10617-10ug

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15924428

  • 205.00€ / 10µg
Expédition estimée: 30-06-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Spécification

Spécification

Flt3-Ligand Protein
10 μg
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
E. coli
TQDCSFQHSP ISSDFAVKIR ELSDYLLQDY PVTVPSNLQD EELCGALWRL VLAQRWMERL KTVAGSKMQG LLERVNTEIH FVTKCAFQHP PSCLRFVQTN ISRLLQETSE QLVALKPWIT RQNFSRCLEL QCQPDSSTLP PPRSPGALEA TALTAPQRP
Greater than 97.0% as determined by SDS-PAGE.
Research Use Only
The ED50 as calculated by the dose-dependant stimulation of the proliferation of human AML5 cells is less than 1.0ng/ml, corresponding to a specific activity of 1,000,000IU/mg.
Rhesus Macaque
Untagged
Lyophilized from a 0.2microm filtered concentrated solution in PBS, pH 7.4.
Vidéos
FDS
Documents
Certificats
Promotions

Promotions