missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Virus Parvovirus B19 VLP VP1/VP2 Protein
A cDNA sequence encoding the Parvovirus B19 VLP VP1/VP2 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP12967-20ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
Virus Parvovirus B19 VLP VP1/VP2 Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
Parvovirus B19 VLP VP1/VP2 protein solution is supplied in 16mM Sodium phosphate pH 7.4, 120mM NaCl and 20% glycerol. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Virus | |
Untagged |