missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Yeast IDH1 Protein
A cDNA sequence encoding the IDH1 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10670-50mg
Dettagli aggiuntivi : Poids : 0.01000kg
Specifica
IDH1 Protein | |
Research Use Only | |
The specific activity was found to be 115 U/mg. | |
Yeast | |
Untagged | |
Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
50 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
Yeast | |
One ml of solution contains 0.075 mol/l KPO4, 50% Glycerol, pH 7.1. |