missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Zika Envelope Protein

A cDNA sequence encoding the Zika Envelope was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP13997-HIS-500ug

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15905369

  • 870.00€ / 500µg
Expédition estimée: 05-07-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Spécification

Spécification

Zika Envelope Protein
Research Use Only
Recombinant Protein
E. coli
MMMASKDATS SVDGASGAGQ LVPEVNASDP LAMDPVAGSS TAVATAGQVN PIDPWIINNF VQAPQGEFTI SPNNTPGDVL FDLSLGPHLN PFLLHLSQMY NGWVGNMRVR IMLAGNAFTA GKIIVSCIPP GFGSHNLTIA QATLFPHVIA DVRTLDPIEV PLEDVRNVLF HNNDRNQQTM RLVCMLYTPL RTGGGTGDSF VVAGRVMTCP SPDFNFLFLV PPTVEQKTRP FTLPNLPLSS LSNSRAPLPI SSMGISPDNV QSVQFQNGRC TLDGRLVGTT PVSLSHVAKI RGTSNGTVIN LTELDGTPFH PFEGPAPIGF PDLGGCDWHI NMTQFGHSSQ TQYDVDTTPD TFVPHLGSIQ ANGIGSGNYV GVLSWISPPS HPSGSQVDLW KIPNYGSSIT EATHLAPSVY PPGFGEVLVF FMSKMPGPGA YNLPCLLPQE YISHLASEQA PTVGEAALLH YVDPDTGRNL GEFKAYPDGF LTCVPNGASS GPQQLPINGV FVFVSWVSRF YQLKPVGTAS SARGRLGLRR
Zika Envelope protein is >95% pure as determined by SDS-PAGE.
500 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Zika
His
Zika Envelope protein solution contains PBS and 25mM arginine.
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions