missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Zika Envelope Protein
A cDNA sequence encoding the Zika Envelope was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP13997-HIS-10ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
Zika Envelope Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MMMASKDATS SVDGASGAGQ LVPEVNASDP LAMDPVAGSS TAVATAGQVN PIDPWIINNF VQAPQGEFTI SPNNTPGDVL FDLSLGPHLN PFLLHLSQMY NGWVGNMRVR IMLAGNAFTA GKIIVSCIPP GFGSHNLTIA QATLFPHVIA DVRTLDPIEV PLEDVRNVLF HNNDRNQQTM RLVCMLYTPL RTGGGTGDSF VVAGRVMTCP SPDFNFLFLV PPTVEQKTRP FTLPNLPLSS LSNSRAPLPI SSMGISPDNV QSVQFQNGRC TLDGRLVGTT PVSLSHVAKI RGTSNGTVIN LTELDGTPFH PFEGPAPIGF PDLGGCDWHI NMTQFGHSSQ TQYDVDTTPD TFVPHLGSIQ ANGIGSGNYV GVLSWISPPS HPSGSQVDLW KIPNYGSSIT EATHLAPSVY PPGFGEVLVF FMSKMPGPGA YNLPCLLPQE YISHLASEQA PTVGEAALLH YVDPDTGRNL GEFKAYPDGF LTCVPNGASS GPQQLPINGV FVFVSWVSRF YQLKPVGTAS SARGRLGLRR | |
Zika Envelope protein is >95% pure as determined by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Zika | |
His | |
Zika Envelope protein solution contains PBS and 25mM arginine. |