Learn More
BTX™ Replacement Pins for Plate Handler
Replacement Pins designed to work with BTX™ Plate Handlers.
Marque: BTX™ 450468
Code nomenclature Nacres: NA.55
Informations supplémentaires : Poids : 0.25000kg
Description
Broches de rechange pour manipulateurs de plaques BTX™
Spécification
High Throughput transfection | |
Gold-plated | |
Épingles de remplacement | |
Pin | |
25 Paquets | |
Banana Cables | |
Yes |
HT 100, HT 200 | |
BTX Electroporation | |
Replacement Pins for Plate Handler, Pkg. of 25 | |
25 x Replacement Pins | |
Non-sterile | |
Exponential Decay Wave, Square Wave |