missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RGS1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Marque: Novus Biologicals NBP2-97095-50ul
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
RGS1 Polyclonal antibody specifically detects RGS1 in Human samples. It is validated for ELISA
Spécification
| RGS1 | |
| Polyclonal | |
| ELISA 1:5000-1:10000 | |
| 1R20, 1R20Early response protein 1R20, B-cell activation protein BL34, BL34, Early response protein 1R20, HEL-S-87, IER1, IR20, IR20immediate-early response 1, B-cell specific, regulator of G-protein signaling 1, regulator of G-protein signalling 1, RGS1 | |
| Produced in rabbits immunized with purified, recombinant Human RGS1 (Accession#: NP_002913.3; Met1-Lys196) | |
| 50 μg | |
| Neuroscience, Neurotransmission, Signal Transduction | |
| 5996 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu