missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Rhesus macaque VEGF R2 / KDR Protein, His Tag

Marque:  ACROBiosystems VE2-C52H3-1MG

Informations supplémentaires : Poids : 0.00001kg

 Afficher plus de versions de ce produit

Code produit. 15877917

  • 2419.31€ / 1mg
Expédition estimée: 27-05-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Kinase insert domain receptor (KDR) is also known as CD309, FLK1, VEGFR, VEGFR2, and is one of the subtypes of VEGFR. VEGF receptors are receptors for vascular endothelial growth factor (VEGF). There are three main subtypes of VEGFR, numbered 1, 2 and 3. The VEGF receptors have an extracellular portion consisting of 7 immunoglobulin-like domains, a single transmembrane spanning region and an intracellular portion containing a split tyrosine-kinase domain. VEGF-A binds to VEGFR-1 (Flt-1) and VEGFR-2 (KDR/Flk-1). VEGFR-2 appears to mediate almost all of the known cellular responses to VEGF.The function of VEGFR-1 is less well defined, although it is thought to modulate VEGFR-2 signaling. Another function of VEGFR-1 may be to act as a dummy/decoy receptor, sequestering VEGF from VEGFR-2 binding (this appears to be particularly important during vasculogenesis in the embryo). In addition, VEGFR2 is able to interact with HIV-1 extracellular Tat protein upon VEGF activation, and seems to enhance angiogenesis in Kaposi's sarcoma lesions.

This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 85.2 kDa. The protein migrates as 120-130 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spécification

Spécification

85.2 kDa
Rhesus macaque VEGF R2, His Tag (VE2-C52H3) is expressed from human 293 cells (HEK293). It contains AA Ala 20 - Glu 764 (Accession # F7E313).
-20°C
VEGF R2
Rhesus Macaque
PBS, pH7.4
HEK293
Bio-Markers and CD Antigens
1 mg
Ala 20 - Glu 764
KDR,CD309,FLK1,VEGFR,VEGFR2
Unconjugated
His Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>97% as determined by SDS-PAGE
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions