missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RhoH Polyclonal Antibody

RhoH Polyclonal Antibody
Brand: Invitrogen PA575334
This item is not returnable.
View return policy
Description
RhoH Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
Specifications
| RhoH | |
| Polyclonal | |
| Unconjugated | |
| RHOH | |
| 5830400A04Rik; ARHH; AU019774; GTP-binding protein TTF; ras homolog family member H; ras homolog gene family, member H; RHOH; rho-related GTP-binding protein RhoH; Translocation three four protein; TTF; TTF, translocation three four | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 305341, 399, 74734 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q15669, Q9D3G9 | |
| RHOH | |
| Synthetic peptide corresponding to amino acids 102-152 of Human RhoH. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction