Learn More
Ribosomal Protein S6/RPS6 Antibody (522731), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Marque:  Novus Biologicals FAB5436MFV500
Code nomenclature Nacres: NA.46
Code produit. 30060754
  • 652.00€ / 0.10mL
Description
Ribosomal Protein S6/RPS6 Monoclonal antibody specifically detects Ribosomal Protein S6/RPS6 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry
TRUSTED_SUSTAINABILITY
Spécification
Spécification
Ribosomal Protein S6/RPS6
Monoclonal
mFluor Violet 500 SE
50mM Sodium Borate
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
Human, Mouse, Rat
Purified
Western Blot, Immunocytochemistry
522731
Western Blot, Immunocytochemistry
Phosphoprotein NP33, 40S ribosomal protein S6, pS6, ribosomal protein S6, S6
E. coli-derived recombinant human RPS6, Met1-Lys249, Accession # P62753
0.1 mL
Cancer, DNA Repair, mTOR Pathway, Phospho Specific, Translation Control
6194
Store at 4C in the dark.
IgG2b