missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNA Polymerase II/POLR2A Antibody (4H8), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Marque: Novus Biologicals NB200-598H
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
RNA Polymerase II/POLR2A Monoclonal specifically detects RNA Polymerase II/POLR2A in Human, Mouse, Yeast, Primate (Negative) samples. It is validated for Western Blot, ELISA.
Spécification
| RNA Polymerase II/POLR2A | |
| Monoclonal | |
| HRP | |
| P24928 | |
| POLR2A | |
| 10 repeats of synthetic peptide YSPTSPS using chemically synthesized phospho-Ser 5 YSPTSpPS (Human). [UniProt# P24928] | |
| Protein G purified | |
| RUO | |
| Primary | |
| Please note that this antibody detects both unphosphorylated and phosphorylated forms of the protein. When using this antibody in western blot and IP, it recognizes both the II0 and IIA form of RPB1 (ie the largest RNAPII subunit). These different forms of RPB1 (as seen in gels) have traditionally been loosely defined as 'hyper-' and 'hypo-phosphorylated', respectively. Here, however, it is important to remember that RPB1 contains no less than 52 repeats of a heptapeptide (TSPTSPS) that can be phosphorylated at different positions (Tyr1, Ser2, Thr4, Ser5 and Ser7; this antibody was raised against a synthetic peptide with 10 of these repeats, mostly but probably not entirely, chemically phosphorylated at Ser5). So, when we say that it recognizes both forms (II0/hyper-phos and IIA/hypo-phos), it might really just mean that the antibody recognizes a few phosphorylated Ser5s among 52 otherwise mostly un-phosphorylated repeats. Alternatively, it might conceivably (though less likely) recognize UN-phosphorylated repeats as there might conceivably be a few of those even in the II0/hyper-phos RPB1 form.' | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot, ELISA | |
| 4H8 | |
| Western Blot, ELISA | |
| DNA-directed RNA polymerase II largest subunit, RNA polymerase II 220 kdsubunit, DNA-directed RNA polymerase II subunit A, DNA-directed RNA polymerase II subunit RPB1, DNA-directed RNA polymerase III largest subunit, EC 2.7.7.48, EC 2.7.7.6, hRPB220, hsRPB1, MGC75453, POLR2, POLRA, polymerase (RNA) II (DNA directed) polypeptide A (220kD), polymerase (RNA) II (DNA directed) polypeptide A, 220kDa, RNA Pol 2, RNA Polymerase 2, RNA polymerase II subunit B1, RNA-directed RNA polymerase II subunit RPB1, RNAPII, RPB1, RPBh1, RpIILS, RPO2, RPOL2 | |
| Mouse | |
| 217 kDa | |
| 0.1 mL | |
| Chromatin Research, Epigenetics, microRNA, Phospho Specific, Transcription Factors and Regulators | |
| 5430 | |
| Human, Mouse, Yeast, Primate (Negative) | |
| Purified |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu