missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ROC1 Recombinant Rabbit Monoclonal Antibody (24GB950)

Rabbit Recombinant Monoclonal Antibody
Marque: Invitrogen™ MA557231
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
ROC1 Recombinant Monoclonal Antibody for Western Blot, Flow
This locus encodes a RING finger-like domain-containing protein. The encoded protein interacts with cullin proteins and likely plays a role in ubiquitination processes necessary for cell cycle progression. This protein may also affect protein turnover. Related pseudogenes exist on chromosomes 2 and 5.
Spécification
| ROC1 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| RBX1 | |
| 1500002P15Rik; AA517855; BA554C12.1; E3 ubiquitin-protein ligase RBX1; E3 ubiquitin-protein ligase RBX1, N-terminally processed; E3 ubiquitin-protein transferase RBX1; E3 ubiquitin-protein transferase RBX1, N-terminally processed; Protein ZYP; Rbx1; Regulator of cullins 1; RING box protein 1; RING finger protein 75; ring-box 1; ring-box 1, E3 ubiquitin protein ligase; RING-box protein 1; RNF75; ROC1; ZYP protein | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 300084, 56438, 9978 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot | |
| 24GB950 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P62877, P62878 | |
| RBX1 | |
| A synthesized peptide derived from human ROC1 (aa 60-108). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu