Learn More
rpmB/L28 Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Marque: Novus Biologicals NBP3-23513PCP
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
rpmB/L28 Polyclonal antibody specifically detects rpmB/L28 in E. coli samples. It is validated for Western Blot, ELISA
Spécification
| rpmB/L28 | |
| Polyclonal | |
| Western Blot, ELISA | |
| RUO | |
| Store at 4C in the dark. |
| Western Blot, ELISA | |
| PerCP | |
| Affinity purified | |
| 946941 | |
| IgG |