missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPS6KA1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
121.38€ - 325.38€
Spécification
| Antigène | RPS6KA1 |
|---|---|
| Concentration | 0.13 mg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugué | Unconjugated |
Description
This gene encodes a zinc finger protein of unknown function. It bears similarity to a zinc finger protein which acts as a transcriptional activator. This gene lies in a region of the X chromosome which has been associated with mental retardation.Spécification
| RPS6KA1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Rat, Mouse | |
| P18653, Q15418, Q63531 | |
| 20111, 6195, 81771 | |
| RPS6KA1 Fusion Protein Ag9537 | |
| Primary | |
| -20°C | |
| RPS6KA1 |
| 0.13 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| HU 1, MAPKAP kinase 1a, MAPKAPK 1a, MAPKAPK1A, p90 RSK 1, p90RSK1, p90S6K, Ribosomal S6 kinase 1, RPS6KA1, RSK, RSK 1, RSK1, S6K alpha 1 | |
| RPS6KA1 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit