Learn More
S-arrestin Antibody (S128), Novus Biologicals™
Mouse Monoclonal Antibody
Marque:  Novus Biologicals NBP2-25161
Code nomenclature Nacres: NA.46
Code produit. 18747043
  • 480.00€ / 0.10mL
Description
S-arrestin Monoclonal antibody specifically detects S-arrestin in Human, Mouse, Rat, Porcine, Bovine, Equine, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen).
TRUSTED_SUSTAINABILITY
Spécification
Spécification
Immunofluorescence
Western Blot 1:5000, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence 1:1000, Immunohistochemistry-Frozen
arrestin 1, RP47,48 kDa protein, S-antigen; retina and pineal gland (arrestin), S-arrestin, visual arrestin
Mouse, Rat, Pig, Bovine
For Research Use Only