missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ S1PR2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marque: Invitrogen™ PA523208
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
S1PR2 Polyclonal Antibody for ICC/IF, Western Blot
This gene encodes a member of the G protein-coupled receptors, as well as the EDG family of proteins. The encoded protein is a receptor for sphingosine 1-phosphate, which participates in cell proliferation, survival, and transcriptional activation. Defects in this gene have been associated with congenital profound deafness.
Spécification
| S1PR2 | |
| Polyclonal | |
| Unconjugated | |
| S1PR2 | |
| agr16; edg5; edg-5; S1PR2 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 14739, 29415, 9294 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide | |
| O95136, P47752, P52592 | |
| S1PR2 | |
| A synthetic peptide corresponding to amino acids 325-343 of human S1P2/Edg5. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu