Learn More
uvex Safety Gloves HexArmor S6
Tous les produits Uvex Produits
Click to view available options
Dimensions:
6
7
8
9
10
11
Conditionnement:
1 paire
Description
HexArmor™ brand products stand for exceptional protection.
- The exclusive SuperFabric™ brand material offers pinprick protection and extremely high cut protection.
- For exceptional impact protection, the patented IR-X™ Impact Exoskeleton™ is an attractive solution.
Spécification
Spécification
| Structure | Seamless Knit |
| Coating Material | Polyurethane |
| Couleur | Blue, Light Blue |
| Cut Resistance Rating | ANSI A6 |
| Hand | Ambidextrous |
| Latex Free | Yes |
| Matériau | SuperFabric™ |
| Type de produit | Cut-resistant Gloves |
| Dimensions | 6 |
| Sexe | Unisex |
| Afficher plus |