missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SARS-CoV-2 ORF3a Antibody, R&D Systems™

Mouse Monoclonal Antibody
Marque: R&D Systems MAB10706-100
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
ORF3a Monoclonal antibody specifically detects ORF3a in SARS-CoV-2 samples. It is validated for Western Blot, Western Blot, Immunocytochemistry
Spécification
| ORF3a | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| SARS-CoV-2 ORF3a peptide | |
| 100 μg | |
| Primary | |
| SARS-CoV-2 | |
| Purified |
| Western Blot, Immunocytochemistry | |
| 1035921 | |
| Western Blot 1 ug/mL, Simple Western 20 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG2a |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu