missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SARS-CoV-2 ORF8 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Marque: Novus Biologicals NBP3-05732
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
SARS-CoV-2 ORF8 Polyclonal antibody specifically detects SARS-CoV-2 ORF8 in SARS-CoV-2 samples. It is validated for Western Blot, ELISA
Spécification
| SARS-CoV-2 ORF8 | |
| Polyclonal | |
| PBS, pH7.2 | |
| Rabbit | |
| Epitope affinity purified | |
| RUO | |
| Primary | |
| SARS-CoV-2 | |
| Purified |
| Western Blot, ELISA | |
| Unconjugated | |
| 2019-nCoV ORF8, 2019-nCoV ORF8 Protein, COVID-19 Non-structural protein 8, COVID-19 ns8, COVID-19 ORF8, Human coronavirus ORF8 Protein, Non-structural protein 8, ns8, ORF8 protein, SARS-CoV-2, SARS-CoV-2 Non-structural protein 8, SARS-CoV-2 ns8, SARSCoV2 ORF8 Protein, SARS-CoV-2 ORF8 Protein, Severe Acute Respiratory Syndrome Coronavirus 2 ORF8 Protein | |
| SARS-CoV-2 ORF8 recombinant protein | |
| 50 μg | |
| Infections (Virus Bacteria and Parasites) | |
| 43740577 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Spot an opportunity for improvement?Share a Content Correction