Learn More
SARS-CoV-2 Spike Human anti-Virus, Unconjugated, Clone: AM002414, Proteintech
Human Recombinant Antibody
Marque: Proteintech 91349-PTG-100UG
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The SARS-CoV-2 Spike antibody from Proteintech is a human recombinant antibody generated with virus of virus SARS-CoV-2 Spike. This antibody recognizes virus antigen. The SARS-CoV-2 Spike antibody has been validated for the following applications: Neutralization, ELISA analysis.
Spécification
| SARS-CoV-2 Spike | |
| Monoclonal | |
| Unconjugated | |
| 100 μg | |
| 43740568 | |
| IgG1 |
| Neutralization, ELISA | |
| AM002414 | |
| Human | |
| Primary | |
| Virus |