missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ SARS-CoV-2 Spike S1 Antibody [PE], Novus Biologicals™
Rabbit Polyclonal Antibody
Marque: Novus Biologicals™ NBP3-26918PE
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
SARS-CoV-2 Spike S1 Polyclonal antibody specifically detects SARS-CoV-2 Spike S1 in SARS-CoV-2 samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunocytochemistry/ Immunofluorescence
Spécification
| SARS-CoV-2 Spike S1 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| SARS-CoV-2 | |
| Purified |
| ELISA, Immunohistochemistry, Western Blot, Immunocytochemistry/Immunofluorescence | |
| PE | |
| SARS-CoV-2 | |
| SARS-CoV-2 Spike S1 Antibody was raised against a peptide corresponding to 16 amino acids near the amino terminus of SARS-CoV-2 (COVID-19) Spike S1 glycoprotein. The immunogen is located within the first 50 amino acids of SARS-CoV-2 (COVID-19) Spike S1 protein. | |
| 0.1 mL | |
| Virology Bacteria and Parasites | |
| 43740568 | |
| Store at 4°C in the dark. | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu