Learn More
Invitrogen™ SBDS Polyclonal Antibody

Rabbit Polyclonal Antibody
Marque: Invitrogen™ PA528950
Description
Recommended positive controls: 293T, A431, H1299, HeLa, HepG2, Molt-4, Raji, mouse liver. Predicted reactivity: Mouse (97%), Rat (97%), Zebrafish (82%), Xenopus laevis (86%), Pig (99%), Chicken (89%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a member of a highly conserved protein family that exists from archaea to vertebrates and plants. The encoded protein may function in RNA metabolism. Mutations within this gene are associated with Shwachman-Bodian-Diamond syndrome. An alternative transcript has been described, but its biological nature has not been determined. This gene has a closely linked pseudogene that is distally located.
Spécification
| SBDS | |
| Polyclonal | |
| Unconjugated | |
| SBDS | |
| 4733401P19Rik; AI836084; CGI-97; FLJ10917; Protein 22A3; ribosome maturation protein SBDS; SBDS; SBDS ribosome assembly guanine nucleotide exchange factor; SBDS ribosome maturation factor; SBDS, ribosome assembly guanine nucleotide exchange factor; SBDS, ribosome maturation factor; SDS; Shwachman-Bodian-Diamond syndrome; Shwachman-Bodian-Diamond syndrome homolog; Shwachman-Bodian-Diamond syndrome homolog (human); Shwachman-Bodian-Diamond syndrome protein; Shwachman-Bodian-Diamond syndrome protein homolog; SWDS; zgc:56700 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 51119, 66711 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 1.07 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| P70122, Q9Y3A5 | |
| SBDS | |
| Full length human SBDS Recombinant protein. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.