Learn More
SBNO1 Antibody, DyLight 550, Novus Biologicals™
Rabbit Polyclonal Antibody
Marque: Novus Biologicals NBP3-05794R
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
SBNO1 Polyclonal specifically detects SBNO1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Spécification
| SBNO1 | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence | |
| FLJ10701, FLJ10833, FLJ14066, FLJ16176, FLJ35199, monocyte protein 3, MOP3, protein strawberry notch homolog 1, Sno, strawberry notch homolog 1 (Drosophila) | |
| Produced in rabbits immunized with E. coli-derived Human SBNO1 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunofluorescence | |
| DyLight 550 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 55206 | |
| Store at 4°C in the dark. | |
| IgG |