Learn More
Sca1 (Ly-6A/E) Rat, anti-Mouse, PE, Clone: E13-161.7, STEMCELL Technologies
Rat Monoclonal Antibody
Marque: Stemcell Technologies 60032PE.1
Description
- The E13-161.7 antibody reacts with Sca1 (Stem Cell Antigen-1 or Ly-6A/E), an 18 kDa GPI-linked protein belonging to the lymphocyte activation protein-6 (Ly-6) family
- Sca1 is expressed on the surface of hematopoietic stem and progenitor cells, myeloid cells, and peripheral B and T lymphocytes
- Sca1 expression levels are strongly upregulated in all strains upon cellular activation
- Sca1 is involved in the regulation of T and B cell responses and is believed to play roles in the differentiation, proliferation, and survival of a variety of stem cells
- Sca1 has emerged as a phenotypic marker of choice for identifying and isolating hematopoietic stem and progenitor cells
Spécification
| Sca1 (Ly-6A/E) | |
| Monoclonal | |
| PE | |
| 50 μg | |
| Mouse | |
| Hematopoietic Stem and Progenitor Cells, Mammary Cells |
| Immunohistochemistry, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Functional Assay | |
| E13-161.7 | |
| Rat | |
| Primary | |
| IgG2a κ |
Le programme Encompass de Fisher Scientific propose des articles qui ne font pas partie de notre portefeuille de distribution. Ces produits n'ont généralement pas de photos ou de descriptions détaillées. Cependant, nous nous engageons à améliorer votre expérience d'achat. Veuillez utiliser le formulaire ci-dessous pour nous faire part de vos commentaires sur le contenu de ce produit.