missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
SCAMP5 Polyclonal antibody specifically detects SCAMP5 in Rat samples. It is validated for Western Blot
Spécification
Spécification
| Antigène | SCAMP5 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugué | Alexa Fluor 594 |
| Formule | 50mM Sodium Borate |
| Alias de gène | DKFZp686L1799, hSCAMP5, MGC24969, Sc5, secretory carrier membrane protein 5, secretory carrier-associated membrane protein 5 |
| Espèces hôtes | Rabbit |
| Immunogène | Synthetic Peptide: C N(217) Q P Q T Q Y S A T P N Y T Y S N(233) |
| Méthode de purification | Immunogen affinity purified |
| Quantité | 0.1 mL |
| Afficher plus |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?